Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Saint-Gobain ALUNDUM™ Extraction Thimbles
Wash, ignite, and use repeatedly
Manufacturer:  Saint-Gobain AN889 7902
 View more versions of this product
Catalog No. 09-660A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Used for all kinds of extractions except strong alkalis
  • ALUNDUM AN889 thimbles (Series 09-660) have medium porosity, retain particles approximately 5µm and larger
  • ALUNDUM AN485 thimbles (Series 09-661) have coarse porosity for faster extraction, less retentivity; retain particles approximately 20µm and larger
  • 34 and 45mm diameter thimbles in each series have round bottoms; all other sizes have flat bottoms
†With round bottoms.
Specifications
Specifications
Flat
2.2cm
3.14 in.
5μm
Extraction Thimble
0.86 in.
Used for all kinds of extractions except strong alkalis
8cm
Medium
Alundum™