Learn More
Saint Gobain Performance Plastics™ ALUNDUM™ Extraction Thimbles
Wash, ignite, and use repeatedly
Manufacturer:  Saint Gobain Performance Plastics™ 7902 AL485
 View more versions of this product
Catalog No. 09-661A
Add to cart
Description
Description
  • Used for all kinds of extractions except strong alkalis
  • ALUNDUM AN889 thimbles (Series 09-660) have medium porosity, retain particles approximately 5µm and larger
  • ALUNDUM AN485 thimbles (Series 09-661) have coarse porosity for faster extraction, less retentivity; retain particles approximately 20µm and larger
  • 34 and 45mm diameter thimbles in each series have round bottoms; all other sizes have flat bottoms
†With round bottoms.
Specifications
Specifications
Flat
2.2cm
3.14 in.
20μm
Extraction Thimble
0.86 in.
Used for all kinds of extractions except strong alkalis
8cm
Coarse
Alundum™