Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Saint-Gobain ALUNDUM™ Filtering Crucibles

For melting metals and vaporizing liquids

$462.00

Specifications

Diameter (Metric) Outer 41mm
Product Type Filtering Crucible
For Use With (Application) For melting metals and vaporizing liquids
Height (Metric) 43mm
View More Specs

Products 1
Catalog Number Mfr. No. Material Porosity Price Quantity & Availability  
Catalog Number Mfr. No. Material Porosity Price Quantity & Availability  
08-230B
View Documents
Saint-Gobain
5205 AL889
Alundum Medium
Case of 12 for $462.00
Add to cart
 
Description

Description

  • Not affected by acids or alkalis
  • Porous for filtering with suction and holding finest precipitates
  • Dry quickly to constant weight
  • Available in medium, made from AN889 ALUNDUM; coarse, made from AN485 mix

Specifications

Specifications

41mm
Filtering Crucible
For melting metals and vaporizing liquids
43mm
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit