Learn More
Saint-Gobain™ Bio-Simplex™ Polypropylene Carboys
Manufacturer:  Saint-Gobain™ DCB40120S
 View more versions of this product
Catalog No. 02-611-209
Add to cart
Specifications
Specifications
±5%
Liquid handling and solution storage for general laboratory, process development and bioproduction applications
1352 oz.
Bottom handles for full hand grip, closed cap and spigot
Carboy
Polypropylene
21 CFR 177, USP Class VI
1352.56 oz., 40L
Wide
40L
1352.56 oz.
40L
Yes
Documents
missing translation for 'provideContentCorrection'
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
missing translation for 'productTitle'