Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Saint-Gobain™ Bio-Simplex™ Polypropylene Carboys

Manufacturer:  Saint-Gobain™ DCB10S3

 View more versions of this product

Catalog No. 02-611-204


  Check Availability
Add to cart

Specifications

Specifications

±5%
Liquid handling and solution storage for general laboratory, process development and bioproduction applications
338 oz.
338.14 oz.
10L
Yes
7.62cm
338.14 oz., 10L
Wide
10L
Carboy
Polypropylene
3 in.
21 CFR 177, USP Class VI
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit