Saint-Gobain™ Chemfluor™ Fluoropolymer Tubing

Ideal for semiconductor processing, biotechnology and pharmaceutical transfer, environmental sample preparation, chromatography, Type I Water, cryogenic, and hazardous chemicals

Manufacturer: Saint-Gobain™ TSPTFE012503150

 View more versions of this product

Catalog No. 14-169-15A

  Check Availability
Add to Cart

    Specifications

    Quantity 1 Each
    For Use With (Application) Chemical transfer and pharmaceutical processes
    Certifications/Compliance FDA Compliant
    Material PTFE,PTFE
    Thickness 0.8mm
    Diameter (English) Inner 0.063 in.
    Thickness (English) Wall 0.03 in.
    Diameter (Metric) Inner 1.6mm
    Diameter (Metric) Outer 3.2mm
    Diameter (English) Outer 0.13 in.

  • Description & Specifications
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit