Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Saint-Gobain Chemfluor Fluoropolymer Tubing
Ideal for semiconductor processing, biotechnology and pharmaceutical transfer, environmental sample preparation, chromatography, Type I Water, cryogenic, and hazardous chemicals
$91.30 - $1820.00
Specifications
Certifications/Compliance | Complies with FDA criteria |
---|---|
For Use With (Application) | Ideal for semiconductor processing, biotechnology and pharmaceutical transfer, environmental sample preparation, chromatography, Type I Water, cryogenic, and hazardous chemicals |
Catalog Number | Mfr. No. | Diameter (English) Inner | Diameter (English) Outer | Material | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (English) Inner | Diameter (English) Outer | Material | Price | Quantity & Availability | |||
14-169-16A
|
Saint-Gobain
TSFE14012503150 |
0.063 in. | 1/8 in. | FEP |
Pack of 50 for $91.30
|
|
14-169-16C
|
Saint-Gobain
TSFE14025004750 |
0.156 in. | 1/4 in. | Fluoropolymer |
Pack of 50 for $256.50
|
|
14-169-100
|
Saint-Gobain
TSFE14025006250 |
0.13 in. | 1/4 in. | Fluoropolymer |
Pack of 50 for $348.00
|
|
14-169-16G
|
Saint-Gobain
TSFE14037506250 |
1/4 in. | 3/8 in. | FEP |
Pack of 50 for $475.50
|
|
14-169-16J
|
Saint-Gobain
TSFE14050006250 |
3/8 in. | 1/2 in. | FEP |
Pack of 50 for $725.00
|
|
14-169-15C
|
Saint-Gobain
TSPTFE025004750 |
0.156 in. | 1/4 in. | PTFE |
Pack of 50 for $246.00
|
|
14-169-17J
|
Saint-Gobain
TSPF35050006250 |
3/8 in. | 1/2 in. | PFA |
Pack of 50 for $1,146.00
|
|
14-169-17D
|
Saint-Gobain
TSPF35025003150 |
0.188 in. | 1/4 in. | PFA |
Pack of 50 for $339.50
|
|
14-169-17P
|
Saint-Gobain
TSPF35075006250 |
3/8 in. | 3/4 in. | Fluoropolymer |
Pack of 50 for $1,586.00
|
|
14-169-17G
|
Saint-Gobain
TSPF35037506250 |
1/4 in. | 0.37 in. | PFA |
Pack of 50 for $813.00
|
|
14-169-15P
|
Saint-Gobain
TSPTFE075006250 |
5/8 in. | 3/4 in. | PTFE |
Pack of 50 for $1,820.00
|
|
14-169-15L
|
Saint-Gobain
TSPTFE062506250 |
1/2 in. | 5/8 in. | PTFE |
Pack of 50 for $1,188.00
|
|
14-169-15A
|
Saint-Gobain
TSPTFE012503150 |
0.063 in. | 0.13 in. | PTFE |
Pack of 50 for $117.10
|
|
14-169-15B
|
Saint-Gobain
TSPTFE025006250 |
0.13 in. | 1/4 in. | PTFE |
Pack of 50 for $271.00
|
|
14-169-15N
|
Saint-Gobain
TSPTFE075003150 |
0.68 in. | 3/4 in. | PTFE |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
14-169-16H
|
Saint-Gobain
TSFE14037503150 |
5/16 in. | 3/8 in. | Fluoropolymer |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
14-169-15K
|
Saint-Gobain
TSPTFE050003150 |
7/16 in. | 1/2 in. | PTFE |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
14-169-17C
|
Saint-Gobain
TSPF35025004750 |
0.156 in. | 1/4 in. | PFA |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
14-169-15E
|
Saint-Gobain
TSPTFE031206250 |
0.188 in. | 5/16 in. | PTFE |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
14-169-15H
|
Saint-Gobain
TSPTFE037503150 |
5/16 in. | 0.37 in. | PTFE |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
14-169-17A
|
Saint-Gobain
TSPF35012503150 |
0.063 in. | 1/8 in. | PFA |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Description
- Contains no additives or plasticizers
- Inert to virtually all chemicals
- Nonprotein binding
Order Info
In packs of 50 ft. (15m)
Compliance
Complies with FDA criteria
Specifications
Complies with FDA criteria | |
Ideal for semiconductor processing, biotechnology and pharmaceutical transfer, environmental sample preparation, chromatography, Type I Water, cryogenic, and hazardous chemicals |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title