Learn More
Saint-Gobain™ Chemfluor™ Fluoropolymer Tubing
Ideal for semiconductor processing, biotechnology and pharmaceutical transfer, environmental sample preparation, chromatography, Type I Water, cryogenic, and hazardous chemicals
Manufacturer:  Saint-Gobain™ TSPTFE025004750
 View more versions of this product
Catalog No. 14-169-15C
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
PTFE
0.156 in.
6.4mm
Ideal for semiconductor processing, biotechnology and pharmaceutical transfer, environmental sample preparation, chromatography, Type I Water, cryogenic, and hazardous chemicals
0.25 in.
4mm
Complies with FDA criteria
236psi