Learn More
Saint-Gobain™ Chemfluor™ Fluoropolymer Tubing
Ideal for semiconductor processing, biotechnology and pharmaceutical transfer, environmental sample preparation, chromatography, Type I Water, cryogenic, and hazardous chemicals
Manufacturer:  Saint-Gobain™ TSPF35075006250
 View more versions of this product
Catalog No. 14-169-17P
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Fluoropolymer
0.63 in.
19mm
Ideal for semiconductor processing, biotechnology and pharmaceutical transfer, environmental sample preparation, chromatography, Type I Water, cryogenic, and hazardous chemicals
0.75 in.
16mm
Complies with FDA criteria
114psi