Learn More
Saint-Gobain™ Chemfluor™ Fluoropolymer Tubing
Ideal for semiconductor processing, biotechnology and pharmaceutical transfer, environmental sample preparation, chromatography, Type I Water, cryogenic, and hazardous chemicals
Manufacturer:  Saint-Gobain™ TSFE14037503150
 View more versions of this product
Catalog No. 14-169-16H
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Fluoropolymer
0.31 in.
9.5mm
Ideal for semiconductor processing, biotechnology and pharmaceutical transfer, environmental sample preparation, chromatography, Type I Water, cryogenic, and hazardous chemicals
0.375 in.
8mm
Complies with FDA criteria
113psi