Saint-Gobain™ Chemfluor™ Fluoropolymer Tubing

Ideal for semiconductor processing, biotechnology and pharmaceutical transfer, environmental sample preparation, chromatography, Type I Water, cryogenic, and hazardous chemicals

Manufacturer: Saint-Gobain™ TSPF35025004750

 View more versions of this product

Catalog No. 14-169-17C

  Check Availability
Add to Cart

    Specifications

    Material PFA
    Certifications/Compliance Complies with FDA criteria
    For Use With (Application) Ideal for semiconductor processing, biotechnology and pharmaceutical transfer, environmental sample preparation, chromatography, Type I Water, cryogenic, and hazardous chemicals
    Diameter (English) Outer 0.25 in.
    Diameter (English) Inner 0.156 in.
    Diameter (Metric) Inner 4mm
    Diameter (Metric) Outer 6.4mm

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit