Learn More
Saint-Gobain™ Chemware™ Bendable PTFE Stirring Rods
Contain fully enclosed steel insert for stiffness, yet can be bent to various shapes for use as powered stirrers
Manufacturer:  Saint-Gobain™ D1069088
 View more versions of this product
Catalog No. 14-518-10D
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Fully enclosed steel insert
0.25 in.
10 in.
PTFE
Each
0.6cm
25mm