Learn More
Saint-Gobain Chemware™ FEP Gas Sampling Bags
5mL thick FEP film for low permeability and maximum sample retention
$53.60 - $107.50
Specifications
Product Type Gas Sampling Bag
For Use With (Application) For gas sample analysis, extracting air samples from exhaust systems
Material FEP (Fluorinated Ethylene Propylene)
Thickness (English) 5 mil
Thickness (Metric) 0.127mm
View More Specs
Products 6
Catalog Number Mfr. No. Dimensions (L x W) Includes Capacity (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Includes Capacity (Metric) Price Quantity & Availability  
10-923-6
View Documents
Saint-Gobain
D107500610
6 x 10 in. (15 x 25cm) Septum valve 1.2L Each for $78.50
Case of 10 EA for $640.50
Add to cart
10-923-1
View Documents
Saint-Gobain
D107500110
6 x 10 in. (15 x 25cm) On/off valve 1.2L Each for $82.50
Case of 10 EA for $681.50
Add to cart
10-923-2
View Documents
Saint-Gobain
D107500210
6 x 6 in. (15 x 15cm) On/off valve 0.5L Each for $101.60
Case of 10 EA for $829.00
Add to cart
10-923-5
View Documents
Saint-Gobain
D107500510
6 x 6 in. (15 x 15cm) Septum valve 0.5L Each for $53.60
Case of 10 EA for $427.50
Add to cart
10-923-3
View Documents
Saint-Gobain
D107500310
12 x 12 in. (30 x 30cm) On/off valve 4.7L Each for $107.50
Case of 10 EA for $860.50
Add to cart
10-923-7
View Documents
Saint-Gobain
D107500710
12 x 12 in. (30 x 30cm) Septum valve 4.7L This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
5mL thick FEP film for low permeability and maximum sample retention
For gas sample analysis, extracting air samples from exhaust systems, monitoring air quality or calibrating instruments in the field.
  • Nonabsorbent, nonreactive to resist most corrosives
  • Heat-sealed on all four sides
  • Second outboard seal not required
Order Info
Available with either a nickel-plated brass ON/OFF valve, which accepts 0.25 in. I.D. flexible tubing, or with a PTFE/silicone septum fitting in a polypropylene housing for use with a hypodermic needle; individually packaged
Specifications
Specifications
Gas Sampling Bag
FEP (Fluorinated Ethylene Propylene)
0.127mm
For gas sample analysis, extracting air samples from exhaust systems
5 mil