Learn More
Saint-Gobain Chemware™ Laboratory Tongs
For handling caustic and harmful materials with care
Manufacturer:  Saint-Gobain D1069290
Catalog No. 15-202-5
Add to cart
Description
Description
  • Made from durable stainless steel
  • For the handling of hot, cold, corrosive or sticky materials
  • Coated with nonstick PTFE
  • Contoured jaws for holding crucibles
  • Will not mar or scratch delicate surfaces
  • Length: 241mm (9.5 in.)
Specifications
Specifications
24cm
Laboratory Tong
Handling caustic and harmful materials with care
9.5 in.
Stainless Steel
Non-stick PTFE
Contoured jaw