Learn More
Saint-Gobain Chemware™ PTFE Boiling Stones
Use these PTFE chips to promote gentle, efficient boiling and help prevent overheating of highly volatile liquids. Saint Gobain Performance Plastics™ Chemware™ PTFE Boiling Stones are not affected by acids, hydrocarbons, ketones, esters, alcohols or alkalis.
Manufacturer:  Saint-Gobain D1069103
Exact match for part number 919120
Catalog No. 09-191-20
Add to cart
Description
Description
Use these PTFE chips to promote gentle, efficient boiling and help prevent overheating of highly volatile liquids. Saint Gobain Performance Plastics™ Chemware™ PTFE Boiling Stones are not affected by acids, hydrocarbons, ketones, esters, alcohols or alkalis.
  • Ideal for continuous extraction methods
  • Chemically inert, and not affected1 by boiling acids, hydrocarbons, ketones, esters, alcohols or alkalis
  • Noncontaminating, show no trace of acidity or alkalinity
  • Will not scratch or abrade glassware
  • Leave no powdery residue
  • 16 oz. (450g) plastic bottles
1At temperatures above 260°C (500°F), a few chemicals at high concentration have been reported reactive to PTFE. These include 80% NaOH or KOH, metal hydrides (such as boranes), aluminum chloride, ammonia and certain amines and imines. Also, slow oxidative attack by 70% nitric acid under pressure at 250°C (480°F) has been observed. Special testing is required when extremes of reducing or oxidizing conditions are approached.
Specifications
Specifications
Boiling stones
Extraction methods, chemically inert and suitable for use with acids, hydrocarbons, ketones, esters, alcohols or alkalis
Boiling Stone
16 oz. (450 g)
PTFE