Learn More
Saint-Gobain Chemware™ PTFE-Coated Forceps
For handling hot or chemically active materials
Manufacturer:  Saint-Gobain D1069288
Catalog No. 10-317-10
Add to cart
Description
Description
  • Nickel-plated steel covered with an extra heavy-duty coating of PTFE
  • Nonstick surface is inert
  • Coating prevents scratching glass and other delicate surfaces
Specifications
Specifications
4.5 in.
Steel, Chrome Plated
Each
114 mm
Forceps
Straight