Saint-Gobain™ Chemware™ PTFE Griffin Beakers

Noncontaminating and inert to highly corrosive reagents such as hydrofluoric acid

Manufacturer: Saint-Gobain™ D1069320

 View more versions of this product

Catalog No. 02-586-1D

 N/A
N/A

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Material PTFE (Polytetrafluoroethylene)
    For Use With (Application) Ideal for digestion
    Height (English) 1.26 in.
    Height (Metric) 3.21cm
    Includes Spout
    Temperature (English) Max. Operating 550°F
    Temperature (Metric) Max. Operating 288°C
    Capacity (Metric) 10mL
    Product Type Beaker

    • Nonwettable, unbreakable and heat resistant
    • Easy to clean
    • Broad spout for easy pouring
    • Can be heated on hotplate up to 288°C (550°F) when proper care is exercised
    • Specially molded bottom allows rapid heat transfer
    • Unlike most PTFE beakers, these will boil water when covered

    Applications:

    Ideal for digestion

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit