Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Saint-Gobain Chemware™ PTFE Griffin Beakers

Noncontaminating and inert to highly corrosive reagents such as hydrofluoric acid

Manufacturer:  Saint-Gobain D1069038

 View more versions of this product

Catalog No. 02-586-1H


Add to cart

Description

Description

  • Nonwettable, unbreakable and heat resistant
  • Easy to clean
  • Broad spout for easy pouring
  • Can be heated on hotplate up to 288°C (550°F) when proper care is exercised
  • Specially molded bottom allows rapid heat transfer
  • Unlike most PTFE beakers, these will boil water when covered

Applications:

Ideal for digestion

Specifications

Specifications

Ideal for digestion
9.23cm
550°F
Beaker
PTFE (Polytetrafluoroethylene)
3.63 in.
Spout
288°C
250mL
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit