Learn More
Saint-Gobain™ Chemware™ PTFE Griffin Beakers
Noncontaminating and inert to highly corrosive reagents such as hydrofluoric acid
Manufacturer:  Saint-Gobain™ D1069039
 View more versions of this product
Catalog No. 02-586-1J
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
PTFE (Polytetrafluoroethylene)
4.21 in.
Spout
288°C
400mL
Ideal for digestion
10.7cm
550°F
13.52 oz.
Beaker