Learn More
Saint-Gobain™ Chemware™ PTFE Griffin Beakers
Noncontaminating and inert to highly corrosive reagents such as hydrofluoric acid
Manufacturer:  Saint-Gobain™ D1069040
 View more versions of this product
Catalog No. 02-586-1K
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
PTFE (Polytetrafluoroethylene)
4.9 in.
Spout
288°C
Beaker
Ideal for digestion
12.45cm
550°F
600mL