Learn More
Saint-Gobain Chemware™ PTFE Solid Balls
Minimize “bumping” in boiling
$57.10 - $234.50
Specifications
Material PTFE
View More Specs
Products 3
Catalog Number Mfr. No. Diameter (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Price Quantity & Availability  
09-191-30A
View Documents
Saint-Gobain
D1069326
3 mm Pack of 100 for $57.10
Add to cart
09-191-30C
View Documents
Saint-Gobain
D1069391
9.5 mm Pack of 100 for $234.50
Add to cart
09-191-30B
View Documents
Saint-Gobain
D1069390
6 mm This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
  • For column packing and as mixing beads
  • Inert to most liquids and vapors
  • Usable to 220°C (428°F)
Specifications
Specifications
PTFE