Learn More
Saint-Gobain Chemware™ PTFE Solid Balls
Minimize “bumping” in boiling
Manufacturer:  Saint-Gobain D1069391
 View more versions of this product
Catalog No. 09-191-30C
Add to cart
Description
Description
  • For column packing and as mixing beads
  • Inert to most liquids and vapors
  • Usable to 220°C (428°F)
Specifications
Specifications
PTFE
9.5mm