Learn More
Saint-Gobain™ Chemware™ PTFE Watch Glasses
Used for weighing, crystallization, and drying
Manufacturer:  Saint-Gobain™ D1069016
 View more versions of this product
Catalog No. 02-610-5J
Add to cart
Description
Description
Used for weighing, crystallization, and drying
Specifications
Specifications
8.01 oz.
8.01 oz., 237mL
15cm
For beakers
237mL
5.9 in.
Ideal for weighing, crystallization and drying, and as beaker covers
PTFE