Learn More
Saint-Gobain™ Chemware™ Tedlar™ PVF Gas Sampling Bags Available on GSA/VA Contract for Federal Government customers only.
Good for use with carbon monoxide, sulfur dioxide, hydrogen sulfide, radon and mercaptans
Manufacturer:  Saint-Gobain™ D107501110
 View more versions of this product
Catalog No. 10-923-11
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
EPA, 40CFR, Part 60, App A Method 18
Carbon monoxide, sulfur dioxide, hydrogen sulfide, radon and mercaptans
12 in.
0.07 in.
12 in.
3.8L
PVDF (Polyvinylidene Fluoride)
12 x 12 in. (30 x 30cm)
On/off valve
Gas Sampling Bag
2mm
128.48 oz.
30cm
30cm