Learn More
Saint-Gobain™ Chemware™ Tedlar™ PVF Gas Sampling Bags
Good for use with carbon monoxide, sulfur dioxide, hydrogen sulfide, radon and mercaptans
Manufacturer:  Saint-Gobain™ D10750145
 View more versions of this product
Catalog No. 10-923-14
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
EPA, 40CFR, Part 60, App A Method 18
Carbon monoxide, sulfur dioxide, hydrogen sulfide, radon and mercaptans
24 in.
0.07 in.
24 in.
37.7L
PVDF (Polyvinylidene Fluoride)
24 x 24 in. (61 x 61cm)
On/off valve
Gas Sampling Bag
2mm
1274.78 oz.
61cm
61cm