Learn More
Saint-Gobain™ Fluoroelastomer Fluran™ Tubing
For use in severe environments where other tubings fail—such as peristaltic pump tubing for extremely corrosive materials, solvent recovery systems, dry cleaning fluid lines, and process monitoring equipment.
Manufacturer:  Saint-Gobain™ AGN00007
 View more versions of this product
Catalog No. 14-170-8A
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Fluran
0.13 in.
0.125 in.
0.125 in.
0.13 in.
For use in severe environments