Learn More
Saint-Gobain Norprene™ Food Process Tubing
$391.00 - $564.50
Specifications
Autoclavable Autoclavable
Material Norprene
View More Specs
Products 8
Catalog Number Mfr. No. Maximum Pressure Thickness (English) Price Quantity & Availability  
Catalog Number Mfr. No. Maximum Pressure Thickness (English) Price Quantity & Availability  
02-590-1G
View Documents
Saint-Gobain
AAL00029
13 psi 0.125 in.
Pack of 50 for $391.00
Add to cart
02-590-1J
View Documents
Saint-Gobain
AAL00046
8 psi 0.125 in.
Pack of 50 for $499.00
Add to cart
02-590-1K
View Documents
Saint-Gobain
AAL00053
7 psi 0.125 in.
Pack of 50 for $564.50
Add to cart
02-590-1H
View Documents
Saint-Gobain
AAL00038
10 psi 0.125 in.
Pack of 50 for $416.50
Add to cart
02-590-1B
View Documents
Saint-Gobain
AAL00007
19 psi 0.0625 in. This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
02-590-1C
View Documents
Saint-Gobain
AAL00012
13 psi 0.0625 in. This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
02-590-1E
View Documents
Saint-Gobain
AAL00022
8 psi 0.0625 in. This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
02-590-1A
View Documents
Saint-Gobain
AAL00003
34 psi 0.0625 in. This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Specifications
Specifications
Autoclavable
Norprene