Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Saint-Gobain Norwell™ The Dipstik™ Universal Drum and Tank Samplers

Samples free-flowing liquids from virtually any source

$966.50 - $2070.00

Specifications

Diameter (English) Outer 1 in.
Diameter (Metric) Outer 2.5cm
Product Type Norwell™ Drum and Tank Sampler
For Use With (Application) For sampling free-flowing liquids in almost any application
View More Specs

Products 4
Catalog Number Mfr. No. Capacity Length (English) Length (Metric) Material Price Quantity & Availability  
Catalog Number Mfr. No. Capacity Length (English) Length (Metric) Material Price Quantity & Availability  
13-417-7
View Documents
Saint-Gobain
D8653274
22 oz., 650mL 84 in. 213cm PTFE Each for $2,070.00
Add to cart
 
13-417-6
View Documents
Saint-Gobain
D8653242
10.2 oz., 300mL 40 in. 101.6cm PTFE Each for $1,643.00
Add to cart
 
13-417-8
View Documents
Saint-Gobain
D8653276
10.2 oz., 300mL 40 in. 101.6cm Polypropylene Case of 12 for $966.50
Add to cart
 
13-417-9
View Documents
Saint-Gobain
D8653275
22 oz., 650mL 84 in. 213cm Polypropylene Case of 12 for $1,314.00
Add to cart
 
Description

Description

Samples free-flowing liquids from virtually any source

Samples free-flowing liquids from virtually any source. Choose between a construction of 100% virgin FEP and PTFE with interchangeable parts to assemble a system for specific applications, or disposable polypropylene unit for simple routine sampling. Both versions feature a safety pouring top with snap-close cap to prevent accidental spills during handling. Cap is attached with a tether to prevent misplacement. A PTFE indicator ring marks fluid levels, as well as immiscible layers within drums and tanks. Push/seal bottom snaps closed after sample collection.

Order Info

Diameter, 1 in. (2.5cm).

Specifications

Specifications

1 in.
2.5cm
Norwell™ Drum and Tank Sampler
For sampling free-flowing liquids in almost any application
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit