Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Saint-Gobain Saint-Gobain™ One-Piece ALUNDUM™ Boat with Handle

For determinations using combustion at high temperatures, metal analysis, carbon content in steel and iron

Manufacturer:  Saint-Gobain 3729 AN788

 View more versions of this product

Catalog No. 07-680B

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

  • Aluminum oxide
  • Resistant to 1430°C (2606°F)
  • Low alkali content

Specifications

Specifications

0.62 in.
3.75 x 0.62 x 0.43 in. (95 x 16 x 11mm)
0.43 in.
3.75 in.
Aluminum Oxide
16mm
For determinations using combustion at high temperatures, metal analysis, carbon content in steel and iron
11mm
95mm
Combustion Boat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit