Learn More
Saint-Gobain™ PharMed™ BPT Tubing Available on GSA/VA Contract for Federal Government customers only.
Ideal for life science applications employing perisaltic pumps
Manufacturer:  Saint-Gobain™ AY242029
 View more versions of this product
Catalog No. 14-170-07
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
PharMed BPT
0.63 in.
0.13 in.
15.9mm
Ideal for life science applications employing peristaltic pumps such as diagnostic test product manufacturing, cell harvest, media process system, vaccine manufacturing, bioreactor process lines, production filtration and fermentation, sterile filling, shear-sensitive fluid transfer
25 ft.
-51° to +135°C
Yes
0.38 in.
9.5mm
Fully tested to ISO 10993 standards. Meets USP Class VI, FDA, and NSF criteria.
Perstaltic Pumps
7.6m
3.2mm