Learn More
Saint-Gobain PharMed BPT Tubing
Ideal for life science applications employing perisaltic pumps
$120.20 - $533.00
Specifications
Certifications/Compliance | Fully tested to ISO 10993 standards. Meets USP Class VI, FDA, and NSF criteria. |
---|---|
Autoclavable | Autoclavable |
Material | PharMed BPT |
For Use With (Application) | Ideal for life science applications employing peristaltic pumps such as diagnostic test product manufacturing, cell harvest, media process system, vaccine manufacturing, bioreactor process lines, production filtration and fermentation, sterile filling, shear-sensitive fluid transfer |
Catalog Number | Mfr. No. | Diameter (English) Inner | Diameter (English) Outer | Thickness (English) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (English) Inner | Diameter (English) Outer | Thickness (English) | Price | Quantity & Availability | |||
14-170-00
|
Saint-Gobain
AY242019 |
1/4 in. | 1/2 in. | 0.13 in. |
Each for $391.50
|
|
14-170-01
|
Saint-Gobain
AY242017 |
1/4 in. | 3/8 in. | 0.063 in. |
Each for $249.50
|
|
14-170-03
|
Saint-Gobain
AY242003 |
0.063 in. | 3/16 in. | 0.063 in. |
Each for $226.50
|
|
14-170-02
|
Saint-Gobain
AY242606 |
0.031 in. | 0.16 in. | 0.063 in. |
Each for $141.50
|
|
14-170-04
|
Saint-Gobain
AY242012 |
0.188 in. | 5/16 in. | 0.063 in. |
Each for $232.00
|
|
14-170-05
|
Saint-Gobain
AY242022 |
5/16 in. | 7/16 in. | 0.063 in. |
Pack of 25 for $281.00
|
|
14-170-06
|
Saint-Gobain
AY242038 |
1/2 in. | 3/4 in. | 0.13 in. |
Each for $533.00
|
|
14-170-09
|
Saint-Gobain
AY242605 |
0.02 in. | 0.145 in. | 0.063 in. |
Each for $120.20
|
|
14-170-13
|
Saint-Gobain
AY242046 |
0.031 in. | 7/8 in. | 0.13 in. |
Each for $514.00
|
|
14-170-15
|
Saint-Gobain
AY242007 |
0.13 in. | 1/4 in. | 0.063 in. |
Each for $176.30
|
|
14-170-14
|
Saint-Gobain
AY242002 |
0.063 in. | 0.13 in. | 0.031 in. |
Each for $153.40
|
|
15-437-50
|
Saint-Gobain
AY242005 |
0.094 in. | 0.2188 in. | 0.0625 in. |
Each for $227.50
|
|
15-437-52
|
Saint-Gobain
AY242027 |
3/8 in. | 1/2 in. | 0.0625 in. |
Each for $293.00
|
|
14-170-07
|
Saint-Gobain
AY242029 |
3/8 in. | 5/8 in. | 0.13 in. |
Each for $349.00
|
|
Description
Ideal for life science applications employing perisaltic pumps
- Nontoxic, nonhemolytic, and nonpyrogenic tubing has long flex life and high biocompatibilityoutlasts silicone tubing up to 30 times longer
- Very good general chemical resistance and excellent acid, alkali, and oxidation resistance
- Opaque to visible and UV light, tubing will protect sensitive fluids
- Withstands temperatures from -51° to +135°C (-60° to +275°F) Withstands repeated clean-in-place and steam-in-place cleaning and sterilization; can be repeatably autoclaved for up to five cycle times without affecting overall service life
- Durometer hardness: Shore A, 64
Applications:
Ideal for life science applications employing peristaltic pumps such as diagnostic test product manufacturing, cell harvest, media process system, vaccine manufacturing, bioreactor process lines, production filtration and fermentation, sterile filling, shear-sensitive fluid transfer
Order Info
In pack of 25 ft. (7.6m)
Compliance
Fully tested to ISO 10993 standards. Meets USP Class VI, FDA, and NSF criteria.
Specifications
Fully tested to ISO 10993 standards. Meets USP Class VI, FDA, and NSF criteria. | |
Autoclavable | |
PharMed BPT | |
Ideal for life science applications employing peristaltic pumps such as diagnostic test product manufacturing, cell harvest, media process system, vaccine manufacturing, bioreactor process lines, production filtration and fermentation, sterile filling, shear-sensitive fluid transfer |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.