Learn More
Saint-Gobain Plastic Sanitary Couplings
Manufacturer:  Saint-Gobain ATP375PPSB
 View more versions of this product
Catalog No. 14-169-119
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Reduces dead space and improves flow.
  • Tapered funnel design provides a steady fluid flow.
  • Smooth inner surface prevents particle entrapment.
  • Seamless construction prevents contamination by eliminating leaks and areas of potential bacterial growth.
  • Material meets USP Class VI requirements and is FDA compliant for food contact.
Specifications
Specifications
Not Autoclavable
For bioreactors and fermentors
Polypropylene
93°C
72°F
Tygopure™ Sanitary Fittings
USP Class VI
Fitting Tygopure 3/8 in. Polypropylene
200°F
200 psi
22°C