Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Saint-Gobain Plastic Sanitary Couplings
Manufacturer: Saint-Gobain ATP500PSSB
Description
- Reduces dead space and improves flow.
- Tapered funnel design provides a steady fluid flow.
- Smooth inner surface prevents particle entrapment.
- Seamless construction prevents contamination by eliminating leaks and areas of potential bacterial growth.
- Material meets USP Class VI requirements and is FDA compliant for food contact.
Specifications
USP Class VI | |
Fitting Tygopure 1/2 in. Polysulfone | |
275°F | |
250 psi | |
22°C |
For bioreactors and fermentors | |
Polysulfone | |
135°C | |
72°F | |
Tygopure™ Sanitary Fittings |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title