Learn More
Saint-Gobain™ Plastic Sanitary Couplings
Manufacturer:  Saint-Gobain™ ATP250PFSB
 View more versions of this product
Catalog No. 14-169-121
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
PVDF
USP Class VI
250psi
22° to 93°C
For bioreactors and fermentors
Tygopure™ Sanitary Fittings
72° to 200°F