Learn More
Saint-Gobain™ Tygon™ Clear Tubing, Formula B-44-4X
Dense bore is smoother than bore of most stainless-steel tubing
Manufacturer:  Saint-Gobain™ AAA00011/DEL
 View more versions of this product
Catalog No. 14-169-37
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Tygon
0.188 in.
6.4mm
50 ft.
165°F
0.25 in.
4.8mm
Meets FDA, 3-A Plastics Standard, and NSF Standard 51 criteria.
25psi
0.8mm