Learn More
Saint-Gobain™ Tygon™ E-1000 Non-DEHP Laboratory Tubing
Manufacturer:  Saint-Gobain™ ADK00036
 View more versions of this product
Catalog No. 14-171-121
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
5/2 in.
1/16 in.
15.9mm
50 ft.
6 psi
1/2 in.
12.7mm
1.6mm
15m