Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Saint-Gobain Tygon™ E-1000 Ultra-Soft Tubing

Non-DEHP Laboratory Tubing

$223.00

Specifications

Diameter (English) Outer 1/2 in.
Diameter (Metric) Outer 12.7 mm
Material Tygon
Overall Length (Metric) 15 m
Overall Length (English) 50 ft.
View More Specs

Products 3
Catalog Number Mfr. No. Diameter (English) Inner Diameter (Metric) Inner Thickness (Metric) Thickness (English) Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (English) Inner Diameter (Metric) Inner Thickness (Metric) Thickness (English) Price Quantity & Availability  
14-179-106
View Documents
Saint-Gobain
ADK00027
3/8 in. 9.5 mm 1.6 mm 0.0625 in. Pack of 50 for $223.00
Add to cart
 
14-179-104
View Documents
Saint-Gobain
ADK00019
1/4 in. 6.4 mm 3.2 mm 0.125 in. This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
14-179-103
View Documents
Saint-Gobain
ADK00017/DEL
1/4 in. 6.4 mm 1.6 mm 0.0625 in. This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

Non-DEHP Laboratory Tubing

Excellent solution to applications requiring complicated laboratory set-ups with sharp radius curves and multiple directional changes.

  • Resists twisting and collapse
  • Stays flexible at temperature as low as -67°F (-55°C)
  • Resistant to corrosive chemicals for laboratory applications
  • Low durometer for use in low-torque pump applications
  • Soft and flexible for lab set-ups
  • Low-temperature resistant for low level cryogenic applications
  • Low durometer for use in low-torque pump applications

  • Synthesis and research set ups
  • Analytical instruments
  • Buffer solutions
  • Peristaltic pumps
  • Vent and drain lines

Order Info

Sold in pack of 50 ft. (15m).

Compliance

Meets FDA requirements for food contact, Europe's EU REACH Food Directives, California Prop 65

Specifications

Specifications

1/2 in.
Tygon
50 ft.
12.7 mm
15 m
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit