Learn More
Saint-Gobain™ Tygon™ Formula 2475 High-Purity Tubing
Environmentally friendly—safe to dispose of through incineration
Manufacturer:  Saint-Gobain™ ACG42064
 View more versions of this product
Catalog No. 14-169-970
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Tygon
1 in.
1 in.
Meets USP Class VI, FDA and NSF specifications. Documented biocompatibility to ISO 10993 standards.
0.19 in.
1.375 in.
0.19 in.
1.375 in.
20psi