Learn More
Saint-Gobain™ Tygon™ Fuel and Lubricant Tubing, Formula F-4040-A
For transporting gasoline, heating oils, kerosene, and glycol-based coolants.
Manufacturer:  Saint-Gobain™ AAG00029
 View more versions of this product
Catalog No. 02-597-1B
Add to cart
Description
Description
For transporting gasoline, heating oils, kerosene, and glycol-based coolants.
  • Resists swelling, cracking and brittleness
  • Compatible with most petroleum-based products
  • Ozone and UV light resistant
  • Minimum extractability safeguards fluids being handled and equipment fluid contacts
  • Not for use with strong acids or alkalis; not to be used in contact with food or beverages
  • Distinctive yellow color
Order Info
Durometer hardness: Shore A, 57. In pack of 50 ft. (15m).
Specifications
Specifications
Yellow
Tygon
50 ft.
For transporting gasoline, heating oils, kerosene, and glycol-based coolants
74°C
3/8 in.
5/8 in.
0.125 in.
35psi
–37°C