Learn More
Saint-Gobain™ Tygon™ Fuel and Lubricant Tubing, Formula F-4040-A
For transporting gasoline, heating oils, kerosene, and glycol-based coolants.
Manufacturer:  Saint-Gobain™ AAG00046
 View more versions of this product
Catalog No. 14-169-8P
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Tygon
0.625 in.
Yellow
50 ft.
-37° to +74°C
0.875 in.
0.125 in.
For transporting gasoline, heating oils, kerosene, and glycol-based coolants
25psi