Learn More
Saint-Gobain Tygon™ High-Purity Pressure Tubing
High-purity pressure tubing with low sorption, flexibility and clarity, available in a single tubing
$483.00 - $766.50
Specifications
Material Tygon
View More Specs
Products 3
Catalog Number Mfr. No. Overall Length (English) Maximum Pressure Price Quantity & Availability  
Catalog Number Mfr. No. Overall Length (English) Maximum Pressure Price Quantity & Availability  
02-593-1A
View Documents
Saint-Gobain
ACX00019
50 ft. 150 psi
Pack of 50 for $483.00
Add to cart
02-593-1B
View Documents
Saint-Gobain
ACX00029
50 ft. 135 psi
Pack of 50 for $766.50
Add to cart
02-593-1F
View Documents
Saint-Gobain
ACX42064
25 ft. 125 psi This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
  • Extremely low sorption to aqueous fluids
  • Plasticizer free
  • Tough braid reinforcement for elevated working pressures
  • Meets USP Class VI and FDA Criteria
Specifications
Specifications
Tygon