Learn More
Saint-Gobain™ Tygon™ High-Purity Pressure Tubing
High-purity pressure tubing with low sorption, flexibility and clarity, available in a single tubing
Manufacturer:  Saint-Gobain™ ACX00019
 View more versions of this product
Catalog No. 02-593-1A
  Check Availability
Add to cart
Description
Description
  • Extremely low sorption to aqueous fluids
  • Plasticizer free
  • Tough braid reinforcement for elevated working pressures
  • Meets USP Class VI and FDA Criteria
Specifications
Specifications
Yes
Tygon
50 ft.
150psi