Saint-Gobain™ Tygon™ ND 100-65 Medical Tubings
For blood contact medical devices where non-DEHP plasticized materials are required
Manufacturer: Saint-Gobain™ ADF00012
 View more versions of this product
Catalog No. 14-171-302
  Check Availability
Add to Cart
    Specifications
    Material Non-DEHP
    Color Clear
    Length (English) 50 in.
    Length (Metric) 15.2m
    Thickness 1/16 in.
    Diameter (English) Outer 5/16 in.
    Diameter (English) Inner 3/16 in.
  • Description & Specifications
Related Products
Product Content Feedback