Learn More
Saint-Gobain™ Tygon™ ND 100-65 Medical Tubings
For blood contact medical devices where non-DEHP plasticized materials are required
Manufacturer:  Saint-Gobain™ ADF00017
 View more versions of this product
Catalog No. 14-171-306
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Non-DEHP
1/4 in.
50 in.
1/16 in.
3/8 in.
Clear
15.2m