Saint-Gobain™ Tygon™ ND 100-65 Medical Tubings
For blood contact medical devices where non-DEHP plasticized materials are required
Manufacturer: Saint-Gobain™ ADF00019
 View more versions of this product
Catalog No. 14-171-308
  Check Availability
Add to Cart
    Specifications
    Diameter (English) Outer 1/2 in.
    Material Non-DEHP
    Color Clear
    Length (English) 50 in.
    Length (Metric) 15.2m
    Thickness 1/8 in.
    Diameter (English) Inner 1/4 in.
  • Description & Specifications
Related Products
Product Content Feedback