Learn More
Saint-Gobain™ Tygon™ Plasticizer-Free Tubing, Formulation 2001
Ideal for use in peristaltic pump systems, soap and detergent dispensing, ink transfer, water purification lines, chemical transfer, and food and beverage applications
Manufacturer:  Saint-Gobain™ AE300003
 View more versions of this product
Catalog No. 14-170-100
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Tygon
0.063 in.
0.063 in.
Compliant with FDA 21 CFR, 177.2600
0.0625 in.
0.188 in.
0.0625 in.
0.188 in.
50 ft.