Learn More
Saint-Gobain™ Tygon™ Plasticizer-Free Tubing, Formulation 2001
Ideal for use in peristaltic pump systems, soap and detergent dispensing, ink transfer, water purification lines, chemical transfer, and food and beverage applications
Manufacturer:  Saint-Gobain™ AE300064
 View more versions of this product
Catalog No. 14-170-109
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Plasticizer/oil -free
1.375 in.
0.187 in.
17.0
Clear
50 ft.
0.187 in.
Superior flexibility
1 in.
1 in.
Compliant with FDA 21 CFR, 177.2600
Ideal for a broad range of demanding applications including peristaltic pump systems, soap and detergent dispensing, ink transfer, water purification lines, food and beverage and chemical transfer.
-108°F to 135°F