Learn More
Saint-Gobain Tygon™ Pressure Tubing with Inner Braid, Formula B-44-4X-I.B.
Tubing wall has reinforced braid to handle long-term continuous pressure up to four times greater than nonreinforced tubing will withstand
Specifications
Certifications/Compliance Meets FDA and 3A Plastics Standard for food and dairy products handling and NSF criteria under Standard 51.
Material Tygon
View More Specs
Products 2
Catalog Number Mfr. No. Description Price Qty Availability  
Catalog Number Mfr. No. Description Price Qty Availability  
14-169-12B
View Documents
Saint-Gobain
ADA00029/DEL
N/A 0.13 in. (3.2mm) thick; 0.63 in. (15.9mm) OD; 0.38 in. (9.5mm) ID x 50 ft. (15.2m) This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
14-169-12A
View Documents
Saint-Gobain
ADA00018/DEL
N/A 0.094 in. (2.4mm) thick; 0.44 in. (11mm) OD; 0.25 in. (6.4mm) ID x 50 ft. (15.2m) This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Odorless, tasteless, and nontoxic
  • Smooth bore won't entrap bacteria
  • Outer surface is smooth, minimizing sanitation problems
  • Also offers visual inspection, flow control, and flexibility
Compliance
Meets FDA and 3A Plastics Standard for food and dairy products handling and NSF criteria under Standard 51.
Specifications
Specifications
Meets FDA and 3A Plastics Standard for food and dairy products handling and NSF criteria under Standard 51.
Tygon