Learn More
Saint-Gobain™ Tygon S3™ B-44-4X I.B. Tubing
Manufacturer:  Saint-Gobain™ ADA05086
 View more versions of this product
Catalog No. 14-171-113
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Phthalate-free plastic
3 in.
95mm
Food, dairy, cosmetics
508mm
3-3/4 in.
76.2mm
Clear
20 in.
40 psi