Saint-Gobain™ Tygon S3™ Clear Tubing, Formula B-44-4X, Non-DEHP
Specially designed to ensure bacterial-free fluid path for food processing applications
Manufacturer: Saint-Gobain™ AAA00018
 View more versions of this product
Catalog No. 14-171-149
  Check Availability
Add to Cart
    Specifications
    Color Clear
    Max. Pressure 1379psi
    Diameter (English) Outer 7/16 in.
    Diameter (English) Inner 1/4 in.
  • Description & Specifications
Related Products
Product Content Feedback